3-Fluoroazetidine hydrochloride, 95%
3-Fluoroazetidine hydrochloride, 95%
3-Fluoroazetidine hydrochloride, 95%
Thermo Scientific Chemicals

3-Fluoroazetidine hydrochloride, 95%

CAS: 617718-46-4 | C3H7ClFN | 111.544 g/mol
数量:
250 mg
500 mg
製品番号(カタログ番号) H59454.MD
または、製品番号H59454-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
仕様
CAS617718-46-4
化学物質名または材質3-Fluoroazetidine hydrochloride
式量111.55
健康被害1H315-H319-H335
健康被害2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
さらに表示
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Eva Van Hende.; Guido Verniest.; Frederik Deroose.; Jan-Willem Thuring.; Gregor Macdonald.; Norbert De Kimpe. Synthesis of 1-Boc-3-fluoroazetidine-3-carboxylic Acid. J. Org. Chem. 2009, 74 (5),2250-2253.
  2. Willem Van Brabandt.; Guido Verniest.; Dirk De Smaele.; Guillaume Duvey.; Norbert De Kimpe. Synthesis of 3-Fluoroazetidines. J. Org. Chem. 2006, 71 (18),7100-7102.