1-Methyl-3-n-octylimidazolium chloride, 97%
1-Methyl-3-n-octylimidazolium chloride, 97%
1-Methyl-3-n-octylimidazolium chloride, 97%
Thermo Scientific Chemicals

1-Methyl-3-n-octylimidazolium chloride, 97%

CAS: 64697-40-1 | C12H23ClN2 | 230.78 g/mol
製品番号(カタログ番号) H59534.18
または、製品番号H59534-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS64697-40-1
IUPAC Name3-methyl-1-octyl-1H-imidazol-3-ium chloride
Molecular FormulaC12H23ClN2
InChI KeyOXFBEEDAZHXDHB-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCCN1C=C[N+](C)=C1
さらに表示
Proton NMR>96.0%
Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Xin Lv.; Weiliang Bao. A β-Keto Ester as a Novel, Efficient, and Versatile Ligand for Copper(I)-Catalyzed C-N, C-O, and C-S Coupling Reactions. J. Org. Chem. 2007, 72, (10), 3863-3867.
  2. Guodong Shen.; Xin Lv.; Weixing Qian.; Weiliang Bao. Cu2O-catalyzed Ullmann-type reaction of vinyl bromides with imidazole and benzimidazole. Tetrahedron Letters. 2008, 49, (29-30), 4556-4559.