6-Methoxypyridine-2-carboxylic acid, 97%
6-Methoxypyridine-2-carboxylic acid, 97%
6-Methoxypyridine-2-carboxylic acid, 97%
Thermo Scientific Chemicals

6-Methoxypyridine-2-carboxylic acid, 97%

CAS: 26893-73-2 | C7H7NO3 | 153.137 g/mol
製品番号(カタログ番号)数量
H59652.03
または、製品番号H59652-03
1 g
製品番号(カタログ番号) H59652.03
または、製品番号H59652-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS26893-73-2
IUPAC Name6-methoxypyridine-2-carboxylic acid
Molecular FormulaC7H7NO3
InChI KeyKSWBODXXZITTPO-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=N1)C(O)=O
さらに表示
Appearance (Color)Pale cream to white
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥96.0%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with
RUO – Research Use Only

General References:

  1. Ellis Vincent Brown.; M. B. Shambhu. Hammick reaction of methoxypyridine-2-carboxylic acids with benzaldehyde. Preparation of methoxy-2-pyridyl phenyl ketones.J. Org. Chem. 1971, 36 (14), 2002-2004.
  2. Edward H. Allen.; Susan K. Laird. Improved preparation of 6-methoxybenzoxazolinone. J. Org. Chem. 1971, 36 (14),2004-2005.