Thermo Scientific Chemicals

Nalpha-Fmoc-O-tert-butyl-L-threonine, 98%, Thermo Scientific Chemicals

Catalog number: H59808.09
10 g, Each
Thermo Scientific Chemicals

Nalpha-Fmoc-O-tert-butyl-L-threonine, 98%, Thermo Scientific Chemicals

Catalog number: H59808.09
10 g, Each
Quantity
Catalog number: H59808.09
also known as H59808-09
Price (USD)
Price: 98.70
Online price: 84.65
Your price:
Quantity
-

Chemical Identifiers

CAS
71989-35-0
IUPAC Name
(2S,3R)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Molecular Formula
C23H27NO5
InChI Key
LZOLWEQBVPVDPR-VLIAUNLRSA-N
SMILES
C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Appearance (Color)
White
Form
Powder
Assay (HPLC)
>97.5%
Optical Rotation
+16.4 ? 1.5? (c=1 in ethyl acetate)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text