2-Chloro-N,N-dimethylbenzamide, 97%, Thermo Scientific Chemicals
2-Chloro-N,N-dimethylbenzamide, 97%, Thermo Scientific Chemicals
2-Chloro-N,N-dimethylbenzamide, 97%, Thermo Scientific Chemicals
2-Chloro-N,N-dimethylbenzamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-N,N-dimethylbenzamide, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H59948.03
または、製品番号H59948-03
1 g
製品番号(カタログ番号) H59948.03
または、製品番号H59948-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6526-67-6
Assay (HPLC)>96.0%
2-Chloro-N,N-dimethylbenzamide is involved in the preparation of repellents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-N,N-dimethylbenzamide is involved in the preparation of repellents.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Sperotto, E.; van Klink, G. P. M.; van Koten, G.; de Vries, J. G. The mechanism of the modified Ullmann reaction. Dalton Trans. 2010, 39 (43), 10338-10351.
  2. Lee, W. S.; Park, K. H.; Yoon, Y. J. N,N-Dimethylamination of Acid Chlorides with Dmf. Synth. Commun. 2000, 30 (23), 4241-4245.