1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane, 97+%
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane, 97+%
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane, 97+%
Thermo Scientific Chemicals

1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane, 97+%

CAS: 528854-34-4 | C22H44P2 | 370.542 g/mol
製品番号(カタログ番号) H60017.03
または、製品番号H60017-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS528854-34-4
IUPAC Name1-{2-[2,5-bis(propan-2-yl)phospholan-1-yl]ethyl}-2,5-bis(propan-2-yl)phospholane
Molecular FormulaC22H44P2
InChI KeyIRVIFEWWPYKALC-UHFFFAOYNA-N
SMILESCC(C)C1CCC(C(C)C)P1CCP1C(CCC1C(C)C)C(C)C
さらに表示
Appearance (Color)White to pale cream
FormPowder or crystalline powder or crystals
Assay (GC)≥96.0%
Proton NMRConforms to structure
It is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep away from strong oxidizing agents.
RUO – Research Use Only