(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%
Thermo Scientific Chemicals

(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%

CAS: 528565-79-9 | C34H36P2 | 506.61 g/mol
製品番号(カタログ番号) H60046.MD
または、製品番号H60046-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS528565-79-9
IUPAC Name(2R,5R)-1-{2-[(2R,5R)-2,5-diphenylphospholan-1-yl]ethyl}-2,5-diphenylphospholane
Molecular FormulaC34H36P2
InChI KeyVHHAZLMVLLIMHT-DXCZPEQUNA-N
SMILESC(CP1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1)P1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)White
Assay from Suppliers CofA≥94%
FormPowder or crystals or crystalline powder
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

Solubility
Sparingly soluble in water.(3.2E-5 g/L) (25°C

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only