2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF
2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF
2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF
Thermo Scientific Chemicals

2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF

CAS: 1053657-14-9 | C8H20NP | 161.23 g/mol
製品番号(カタログ番号) H60435.14
または、製品番号H60435-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
仕様
CAS1053657-14-9
化学物質名または材質2-(Diisopropylphosphino)ethylamine
Concentration or Composition (by Analyte or Components)10% w/w soln. in THF
DOT情報Hazard Class: 3; Packaging Group: III
式量161.22
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only