Dicumyl peroxide, 98%
Dicumyl peroxide, 98%
Dicumyl peroxide, 98%
Dicumyl peroxide, 98%
Thermo Scientific Chemicals

Dicumyl peroxide, 98%

CAS: 80-43-3 | C18H22O2 | 270.372 g/mol
製品番号(カタログ番号) H60442.22
または、製品番号H60442-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS80-43-3
IUPAC Name{2-[(2-phenylpropan-2-yl)peroxy]propan-2-yl}benzene
Molecular FormulaC18H22O2
InChI KeyXMNIXWIUMCBBBL-UHFFFAOYSA-N
SMILESCC(C)(OOC(C)(C)C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)White to pale cream
Assay from Suppliers CofA≥97.5% (HPLC)
FormPowder or crystalline powder or crystals

Dicumyl peroxide (DCP) is used:

  • in vulcanization of rubber.
  • as a cross linking agent in the synthesis of polylactic acid composite fibers.
  • in the preparation of polyethylene composites.
  • in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicumyl peroxide (DCP) is used: • in vulcanization of rubber. • as a cross linking agent in the synthesis of polylactic acid composite fibers. • in the preparation of polyethylene composites. • in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.

Solubility
Soluble in chloroform and most organic solvents. Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents and acids.
RUO – Research Use Only

General References:

  1. Ray A. Dickie.; John D. Ferry. Dynamic Mechanical Properties of Cross-Linked Rubbers. III. Dicumyl Peroxide Vulcanizates of Natural Rubber1. J. Phys. Chem. 1966, 70 (8),2594-2600.
  2. S. Sapieha.; P. Allard.; Y. H. Zang. Dicumyl peroxide-modified cellulose/LLDPE composites. J. Appl. Polym. Sci. 1990, 41 (9-10),2039-2048.