(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene, 97+%
(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene, 97+%
(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene, 97+%
Thermo Scientific Chemicals

(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene, 97+%

CAS: 136705-64-1 | C22H36P2 | 362.478 g/mol
製品番号(カタログ番号) H60470.MD
または、製品番号H60470-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン4814174
CAS136705-64-1
化学物質名または材質(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene
密度1.01
引火点>110°C (230°F)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only