2,3,6-Trimethylphenol, 95%
2,3,6-Trimethylphenol, 95%
2,3,6-Trimethylphenol, 95%
Thermo Scientific Chemicals

2,3,6-Trimethylphenol, 95%

CAS: 2416-94-6 | C9H12O | 136.19 g/mol
製品番号(カタログ番号) H60489.14
または、製品番号H60489-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2416-94-6
Assay (HPLC)>94.0%
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
RUO – Research Use Only

General References:

  1. O.A. Kholdeeva; A.V. Golovin; R.I. Maksimovskaya; I.V. Kozhevnikov. Oxidation of 2,3,6-trimethylphenol in the presence of molybdovanadophosphoric heteropoly acids. Journal of Molecular Catalysis.1992, 75 235-244.
  2. Katsuomi Takehira; Masao Shimizu; Yoshihito Watanabe; Hideo Orita; Takashi Hayakawa. A novel oxygenation of 2,3,6-trimethylphenol to trimethyl-p-benzoquinone by dioxygen with copper(II) chloride/amine hydrochloride catalyst. Tetrahedron Letters.1989, 30 6691-6692.