(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, 97+%
(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, 97+%
(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, 97+%
Thermo Scientific Chemicals

(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine, 97+%

CAS: 960128-64-7 | C22H22NP | 331.40 g/mol
製品番号(カタログ番号) H60538.03
または、製品番号H60538-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS960128-64-7
化学物質名または材質(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine
DOT情報Hazard Class: 9; Packaging Group: III
式量331.39
健康被害1H315-H319-H335
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only