Thermo Scientific Chemicals

n-Octadecyltrimethylammonium chloride, 95%, Thermo Scientific Chemicals

Catalog number: H60665.09
10 g, Each
Thermo Scientific Chemicals

n-Octadecyltrimethylammonium chloride, 95%, Thermo Scientific Chemicals

Catalog number: H60665.09
10 g, Each
Quantity
Catalog number: H60665.09
also known as H60665-09
Price (USD)
Price: 152.00
Online price: 129.65
Your price:
Quantity
-

Chemical Identifiers

CAS
112-03-8
Assay (HPLC)
>94.0%

Description

Emulsion (main components are high polymeric and cyclic silicones); forms a protective film to coat split ends; conditioning agent for rinses and shampoos; glazing agent for hair.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Emulsion (main components are high polymeric and cyclic silicones); forms a protective film to coat split ends; conditioning agent for rinses and shampoos; glazing agent for hair.

Solubility
Soluble in water 1.759 mg/L @ 25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text