Piperonyl butoxide, tech. 90%
Piperonyl butoxide, tech. 90%
Piperonyl butoxide, tech. 90%
Piperonyl butoxide, tech. 90%
Thermo Scientific Chemicals

Piperonyl butoxide, tech. 90%

CAS: 51-03-6 | C19H30O5 | 338.44 g/mol
製品番号(カタログ番号) H60771.AE
または、製品番号H60771-AE
価格(JPY)
-
見積もりを依頼する
数量:
100 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS51-03-6
IUPAC Name5-{[2-(2-butoxyethoxy)ethoxy]methyl}-6-propyl-2H-1,3-benzodioxole
Molecular FormulaC19H30O5
InChI KeyFIPWRIJSWJWJAI-UHFFFAOYSA-N
SMILESCCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC
さらに表示
Assay (GC)>88.0%
Piperonyl butoxide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonyl butoxide is used as a pharmaceutical intermediate.

Solubility
Soluble in benzene, ethanol and methanol. Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Shozo Kuwatsuka.; J. E. Casida. Synergist Synthesis, Synthesis of Methylene-C14-Dioxyphenyl Compounds: Radioactive Safrole, Dihydrosafrole, Myristicin, Piperonyl Butoxide and Diastereoisomers of Sulfoxide. J. Agric. Food Chem. 1965, 13 (6), 528-533.
  2. Claude H. Schmidt.; Paul A. Dahm. Fomina. The synthesis of C14-labeled piperonyl butoxide and its fate in the Madeira roach. Journal of Economic Entomology. 1956, 49 (6), 729-735.