(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone, 98%
(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone, 98%
(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone, 98%
Thermo Scientific Chemicals

(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone, 98%

CAS: 100516-54-9 | C24H21NO4 | 387.44 g/mol
製品番号(カタログ番号)数量
H60821.03
または、製品番号H60821-03
1 g
製品番号(カタログ番号) H60821.03
または、製品番号H60821-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS100516-54-9
IUPAC Namebenzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Molecular FormulaC24H21NO4
InChI KeyHECRUWTZAMPQOS-UHFFFAOYNA-N
SMILESO=C(OCC1=CC=CC=C1)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Enantiomeric excess>98%
FormSolid
Appearance (Color)Off-white
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only