1,3,5-Triethylbenzene, 95%
1,3,5-Triethylbenzene, 95%
1,3,5-Triethylbenzene, 95%
Thermo Scientific Chemicals

1,3,5-Triethylbenzene, 95%

CAS: 102-25-0 | C12H18 | 162.28 g/mol
製品番号(カタログ番号) H60875.14
または、製品番号H60875-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS102-25-0
IUPAC Name1,3,5-triethylbenzene
Molecular FormulaC12H18
InChI KeyWJYMPXJVHNDZHD-UHFFFAOYSA-N
SMILESCCC1=CC(CC)=CC(CC)=C1
さらに表示
Assay (GC)>94.0%
,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.

Solubility
Insoluble in water. Soluble in alcohol, ether.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
RUO – Research Use Only

General References:

  1. W. V. Steele.; R. D. Chirico.; S. E. Knipmeyer.; A. Nguyen. Vapor Pressure, Heat Capacity, and Density along the Saturation Line, Measurements for Dimethyl Isophthalate, Dimethyl Carbonate, 1,3,5-Triethylbenzene, Pentafluorophenol, 4-tert-Butylcatechol, α-Methylstyrene, and N,N‘-Bis(2-hydroxyethyl)ethylenediamine. J. Chem. Eng. Data. 1997, 42 (6),.
  2. Hiromi Ohi.; Yoshimitsu Tachi.; Takashi Kunimoto.; Shinobu Itoh. Structure and photoluminescence property of two-dimensional coordination polymer complexes involving CuI6X6 (X = Cl, Br, I) hexagon prism cluster supported by a tripodal tripyridine ligand with 1,3,5-triethylbenzene spacer. Dalton Trans. 2005, (19),3146-3147.