(R)-2-(Boc-amino)-4-phenylbutyric acid, 98%
(R)-2-(Boc-amino)-4-phenylbutyric acid, 98%
(R)-2-(Boc-amino)-4-phenylbutyric acid, 98%
(R)-2-(Boc-amino)-4-phenylbutyric acid, 98%
Thermo Scientific Chemicals

(R)-2-(Boc-amino)-4-phenylbutyric acid, 98%

CAS: 82732-07-8 | C15H21NO4 | 279.336 g/mol
製品番号(カタログ番号) H61183.03
または、製品番号H61183-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS82732-07-8
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-4-phenylbutanoic acid
Molecular FormulaC15H21NO4
InChI KeyMCODLPJUFHPVQP-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(O)=O
さらに表示
Assay (HPLC)>97.5%
(R)-2-(Boc-amino)-4-phenylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-2-(Boc-amino)-4-phenylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only