Thermo Scientific Chemicals

4-Morpholineacetic acid hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H61188.06
5 g, Each
Thermo Scientific Chemicals

4-Morpholineacetic acid hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H61188.06
5 g, Each
Quantity
Catalog number: H61188.06
also known as H61188-06
Price (USD)
Price: 252.00
Online price: 217.65
Your price:
Quantity
-

Chemical Identifiers

CAS
89531-58-8
IUPAC Name
hydrogen 2-(morpholin-4-yl)acetic acid chloride
Molecular Formula
C6H12ClNO3
InChI Key
ICGYBJNONRDZOP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].OC(=O)CN1CCOCC1
Assay (GC)
>94.0%

Description

4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text