Thermo Scientific Chemicals

4-Amino-5-bromo-2-chloropyrimidine, 95%, Thermo Scientific Chemicals

Catalog number: H61429.14
25 g, Each
Thermo Scientific Chemicals

4-Amino-5-bromo-2-chloropyrimidine, 95%, Thermo Scientific Chemicals

Catalog number: H61429.14
25 g, Each
Quantity
Catalog number: H61429.14
also known as H61429-14
Price (USD)
Price: 590.00
Online price: 510.65
Your price:
Quantity
-

Chemical Identifiers

CAS
205672-25-9
Assay (HPLC)
>94.0%

Description

4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2-4°C. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text