2-Bromo-4-fluoro-1-nitrobenzene, 98%
2-Bromo-4-fluoro-1-nitrobenzene, 98%
2-Bromo-4-fluoro-1-nitrobenzene, 98%
Thermo Scientific Chemicals

2-Bromo-4-fluoro-1-nitrobenzene, 98%

CAS: 700-36-7 | C6H3BrFNO2 | 219.997 g/mol
製品番号(カタログ番号) H61466.14
または、製品番号H61466-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS700-36-7
IUPAC Name2-bromo-4-fluoro-1-nitrobenzene
Molecular FormulaC6H3BrFNO2
InChI KeyVGYVBEJDXIPSDL-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=C(Br)C=C(F)C=C1
さらに表示
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only