Nepsilon-Boc-D-lysine, 98%
Nepsilon-Boc-D-lysine, 98%
Nepsilon-Boc-D-lysine, 98%
Thermo Scientific Chemicals

Nepsilon-Boc-D-lysine, 98%

CAS: 31202-69-4 | C11H22N2O4 | 246.31 g/mol
製品番号(カタログ番号) H62736.03
または、製品番号H62736-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS31202-69-4
IUPAC Name(2R)-2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Molecular FormulaC11H22N2O4
InChI KeyVVQIIIAZJXTLRE-SVGMAFHSNA-N
SMILESCC(C)(C)OC(=O)NCCCC[C@@H](N)C(O)=O
さらに表示
Appearance (Color)White to pale cream
FormPowder
Assay from Supplier's CofA≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only