Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H62975.14
25 g, Each
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H62975.14
25 g, Each
Quantity
Catalog number: H62975.14
also known as H62975-14
Price (USD)
Price: 182.00
Online price: 156.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3705-42-8
IUPAC Name
5-(benzyloxy)-4-{[(benzyloxy)carbonyl]amino}-5-oxopentanoic acid
Molecular Formula
C20H21NO6
InChI Key
VWHKODOUMSMUAF-UHFFFAOYNA-N
SMILES
OC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Assay (HPLC)
>94.0%

Description

N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.

Solubility
Soluble in methanol, dimethyl sulfoxide. Slightly soluble in water.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text