L-Cystine dimethyl ester dihydrochloride, 98%
L-Cystine dimethyl ester dihydrochloride, 98%
L-Cystine dimethyl ester dihydrochloride, 98%
Thermo Scientific Chemicals

L-Cystine dimethyl ester dihydrochloride, 98%

CAS: 32854-09-4 | C8H18Cl2N2O4S2 | 341.26 g/mol
製品番号(カタログ番号) H63234.14
または、製品番号H63234-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS32854-09-4
IUPAC Namedihydrogen methyl 2-amino-3-[(2-amino-3-methoxy-3-oxopropyl)disulfanyl]propanoate dichloride
Molecular FormulaC8H18Cl2N2O4S2
InChI KeyQKWGUPFPCRKKMQ-UHFFFAOYNA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC(=O)C(N)CSSCC(N)C(=O)OC
さらに表示
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only