(S)-2-(Boc-amino)butyric acid, 95%
(S)-2-(Boc-amino)butyric acid, 95%
(S)-2-(Boc-amino)butyric acid, 95%
(S)-2-(Boc-amino)butyric acid, 95%
Thermo Scientific Chemicals

(S)-2-(Boc-amino)butyric acid, 95%

CAS: 34306-42-8 | C9H17NO4 | 203.24 g/mol
製品番号(カタログ番号) H63686.14
または、製品番号H63686-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS34306-42-8
Assay (HPLC)>94.0%
It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Dieter Seebach; Albert K. Beck; Hans G. Bossler; Christian Gerber; Soo Y Ko; C. William Murtiashaw; Reto Naef; Shin-Ichiro Shoda; drian Thaler; Manfred Krieger; and Roland Wenger. Modification of Cyclosporin A (CS): Generation of an enolate at the sarcosine residue and reactions with electrophiles. Helvetica Chimica Acta. 1993, 76(4), 1564-1590.
  2. Zsolt Bozsó; Gergely Tóth; Richard F. Murphy; Sándor Lovas. Difficulties in coupling to conformationally constrained aromatic amino acids. Letters in Peptide Science. 2000, 7(3), 157-163.