1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%
1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%
1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%
Thermo Scientific Chemicals

1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%

CAS: 346451-15-8 | C30H30Cl4N2 | 560.384 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) H63961.MD
または、製品番号H63961-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
仕様
CAS346451-15-8
化学物質名または材質1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride
White
式量560.38
IUPAC名1,4-bis[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only