1H-Indazole-3-carboxylic acid, 98%
1H-Indazole-3-carboxylic acid, 98%
1H-Indazole-3-carboxylic acid, 98%
Thermo Scientific Chemicals

1H-Indazole-3-carboxylic acid, 98%

CAS: 4498-67-3 | C8H6N2O2 | 162.148 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) H64001.06
または、製品番号H64001-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4498-67-3
IUPAC Name1H-indazole-3-carboxylic acid
Molecular FormulaC8H6N2O2
InChI KeyBHXVYTQDWMQVBI-UHFFFAOYSA-N
SMILESOC(=O)C1=NNC2=CC=CC=C12
さらに表示
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only