2-(Trifluoromethyl)pyridine-5-boronic acid, 98%
2-(Trifluoromethyl)pyridine-5-boronic acid, 98%
2-(Trifluoromethyl)pyridine-5-boronic acid, 98%
Thermo Scientific Chemicals

2-(Trifluoromethyl)pyridine-5-boronic acid, 98%

CAS: 868662-36-6 | C6H5BF3NO2 | 190.916 g/mol
製品番号(カタログ番号) H64106.06
または、製品番号H64106-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS868662-36-6
IUPAC Name[6-(trifluoromethyl)pyridin-3-yl]boronic acid
Molecular FormulaC6H5BF3NO2
InChI KeyBNTIPMNMTIAWIW-UHFFFAOYSA-N
SMILESOB(O)C1=CN=C(C=C1)C(F)(F)F
さらに表示
Assay (HPLC)>97.5%
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only