6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride, 98%
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride, 98%
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride, 98%
Thermo Scientific Chemicals

6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride, 98%

CAS: 20232-39-7 | C11H14ClNO2 | 227.69 g/mol
製品番号(カタログ番号)数量
H64130.06
または、製品番号H64130-06
5 g
製品番号(カタログ番号) H64130.06
または、製品番号H64130-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS20232-39-7
IUPAC Name6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride
Molecular FormulaC11H14ClNO2
InChI KeyPQXVEYYRJHMTEV-UHFFFAOYSA-N
SMILESCl.COC1=CC2=C(C=NCC2)C=C1OC
さらに表示
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only