9,9'-Spirobifluorene, 98%
9,9'-Spirobifluorene, 98%
9,9'-Spirobifluorene, 98%
Thermo Scientific Chemicals

9,9'-Spirobifluorene, 98%

CAS: 159-66-0 | C25H16 | 316.403 g/mol
製品番号(カタログ番号) H64133.03
または、製品番号H64133-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS159-66-0
IUPAC Name9,9'-spirobi[fluorene]
Molecular FormulaC25H16
InChI KeySNFCXVRWFNAHQX-UHFFFAOYSA-N
SMILESC1=CC=C2C(=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12
さらに表示
Assay (HPLC)>97.5%
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

Solubility
Soluble in common organic solvents. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. S. Tang; M. R. liu; P. Lu; H. xia; M. li; Z. Q. xie; F. Z. shen; C. Gu; H. P. wang; B. yang; and Y. G. ma. A Molecular Glass for Deep-Blue Organic Light-Emitting Diodes Comprising a 9,9'-Spirobifluorene Core and Peripheral Carbazole Groups. Advanced Functional Materials. 200717(15), 2869-2877.
  2. Jean-Hugues Fournier; Thierry Maris; and James D. Wuest. Molecular Tectonics. Porous Hydrogen-Bonded Networks Built from Derivatives of 9,9‘-Spirobifluorene. J. Org. Chem. 200469(6),1762-1775.