2-Bromo-6-fluoro-4-methylpyridine, 98%
2-Bromo-6-fluoro-4-methylpyridine, 98%
2-Bromo-6-fluoro-4-methylpyridine, 98%
2-Bromo-6-fluoro-4-methylpyridine, 98%
Thermo Scientific Chemicals

2-Bromo-6-fluoro-4-methylpyridine, 98%

CAS: 180608-37-1 | C6H5BrFN | 190.015 g/mol
製品番号(カタログ番号) H64449.06
または、製品番号H64449-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS180608-37-1
IUPAC Name2-bromo-6-fluoro-4-methylpyridine
Molecular FormulaC6H5BrFN
InChI KeyCIDWWTCSJWQJQR-UHFFFAOYSA-N
SMILESCC1=CC(F)=NC(Br)=C1
さらに表示
Assay (HPLC)>97.5%
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only