Thermo Scientific Chemicals

7-Deazahypoxanthine, 97%, Thermo Scientific Chemicals

Catalog number: H64537.06
5 g, Each
Thermo Scientific Chemicals

7-Deazahypoxanthine, 97%, Thermo Scientific Chemicals

Catalog number: H64537.06
5 g, Each
Quantity
Catalog number: H64537.06
also known as H64537-06
Price (USD)
Price: 72.60
Online price: 61.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3680-71-5
IUPAC Name
1H,4H,7H-pyrrolo[2,3-d]pyrimidin-4-one
Molecular Formula
C6H5N3O
InChI Key
FBMZEITWVNHWJW-UHFFFAOYSA-N
SMILES
O=C1N=CNC2=C1C=CN2
Assay (HPLC)
>96.0%

Description

It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Moisture and light sensitive. Store at -20°C
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text