2,5-Dihydroxyterephthalic acid, 97%
2,5-Dihydroxyterephthalic acid, 97%
2,5-Dihydroxyterephthalic acid, 97%
2,5-Dihydroxyterephthalic acid, 97%
Thermo Scientific Chemicals

2,5-Dihydroxyterephthalic acid, 97%

CAS: 610-92-4 | C8H6O6 | 198.13 g/mol
製品番号(カタログ番号)数量
H64888.1425 g
製品番号(カタログ番号) H64888.14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS610-92-4
IUPAC Name2,5-dihydroxybenzene-1,4-dicarboxylic acid
Molecular FormulaC8H6O6
InChI KeyOYFRNYNHAZOYNF-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=C(C=C1O)C(O)=O
さらに表示
Appearance (Color)Yellow
FormPowder
Assay (HPLC)≥96.0%
Identification (FTIR)conforms
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in hot dimethylformamide.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Zhang T, et al. Improvement of surface wettability and interfacial adhesion ability of poly(p-phenylene benzobisoxazole) (PBO) fiber by incorporation of 2,5-dihydroxyterephthalic acid (DHTA). Eur. Polymer J.2011 45(1), 302--307.
  2. Ju HK, et al.Effect of steam treatment on soluble phenolic content and antioxidant activity of the Chaga mushroom (Inonotus obliquus). Food Chem.2010 119(2), 619-625.