ITIC-M, Thermo Scientific Chemicals
ITIC-M, Thermo Scientific Chemicals
ITIC-M, Thermo Scientific Chemicals
ITIC-M, Thermo Scientific Chemicals
Thermo Scientific Chemicals

ITIC-M, Thermo Scientific Chemicals

数量:
100 mg
250 mg
500 mg
製品番号(カタログ番号) H66035.MC
または、製品番号H66035-MC
価格(JPY)
-
見積もりを依頼する
数量:
100 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2047352-80-5
FormFlake or threadlike
Bandgap energy1.65 ? 0.02 eV
Appearance (Color)Black
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only