8-Bromo-2-chloro-6-fluoroquinazoline, 95%, Thermo Scientific Chemicals
8-Bromo-2-chloro-6-fluoroquinazoline, 95%, Thermo Scientific Chemicals
8-Bromo-2-chloro-6-fluoroquinazoline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Bromo-2-chloro-6-fluoroquinazoline, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号) H66128.MD
または、製品番号H66128-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS953039-63-9
IUPAC Name8-bromo-2-chloro-6-fluoroquinazoline
Molecular FormulaC8H3BrClFN2
InChI KeyXTEWVYPXKPYPRE-UHFFFAOYSA-N
SMILESFC1=CC(Br)=C2N=C(Cl)N=CC2=C1
さらに表示
Assay (HPLC)99.40%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only