4-Phenylpiperidine, 96%
4-Phenylpiperidine, 96%
4-Phenylpiperidine, 96%
4-Phenylpiperidine, 96%
Thermo Scientific Chemicals

4-Phenylpiperidine, 96%

CAS: 771-99-3 | C11H15N | 161.248 g/mol
製品番号(カタログ番号) H66389.06
または、製品番号H66389-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン124508
CAS771-99-3
化学物質名または材質4-Phenylpiperidine
色Yellow to Brown
EINECS番号212-243-1
さらに表示
4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
G H Loew; J A Lawson; E T Uyeno; L Toll; G Frenking; W Polgar; L Y Ma; N Camerman; A Camerman. Structure-activity studies of morphine fragments. I. 4-alkyl-4-(m-hydroxy-phenyl)-piperidines. Molecular Pharmacology.1988, 34, (3), 363-376.