N-Boc-L-tryptophan methyl ester, 96%
N-Boc-L-tryptophan methyl ester, 96%
N-Boc-L-tryptophan methyl ester, 96%
Thermo Scientific Chemicals

N-Boc-L-tryptophan methyl ester, 96%

CAS: 33900-28-6 | C17H22N2O4 | 318.37 g/mol
製品番号(カタログ番号) H66442.14
または、製品番号H66442-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS33900-28-6
IUPAC Namemethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate
Molecular FormulaC17H22N2O4
InChI KeyQXLOVPXUZAOKBL-AWEZNQCLSA-N
SMILESCOC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OC(C)(C)C
さらに表示
Appearance (Color)White to pale cream
Assay (HPLC)≥95.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only