4-Chloroindole-3-acetic acid, 95%
4-Chloroindole-3-acetic acid, 95%
4-Chloroindole-3-acetic acid, 95%
Thermo Scientific Chemicals

4-Chloroindole-3-acetic acid, 95%

CAS: 2519-61-1 | C10H8ClNO2 | 209.629 g/mol
製品番号(カタログ番号) H66637.03
または、製品番号H66637-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2519-61-1
IUPAC Name2-(4-chloro-1H-indol-3-yl)acetic acid
Molecular FormulaC10H8ClNO2
InChI KeyWNCFBCKZRJDRKZ-UHFFFAOYSA-N
SMILESOC(=O)CC1=CNC2=CC=CC(Cl)=C12
さらに表示
Appearance (Color)Cream
Assay from Supplier's CofA≥94.0% (GC)
FormPowder or crystals or crystalline powder
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only