alpha-Bromo-4-fluorophenylacetic acid, 96%
alpha-Bromo-4-fluorophenylacetic acid, 96%
alpha-Bromo-4-fluorophenylacetic acid, 96%
alpha-Bromo-4-fluorophenylacetic acid, 96%
Thermo Scientific Chemicals

alpha-Bromo-4-fluorophenylacetic acid, 96%

CAS: 29270-33-5 | C8H6BrFO2 | 233.036 g/mol
製品番号(カタログ番号) H66692.06
または、製品番号H66692-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
仕様
CAS29270-33-5
化学物質名または材質alpha-Bromo-4-fluorophenylacetic acid
White to Brown
DOT情報Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
式量233.03
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only