P3HT (OPV grade - 91-94% RR)
P3HT (OPV grade - 91-94% RR)
P3HT (OPV grade - 91-94% RR)
Thermo Scientific Chemicals

P3HT (OPV grade - 91-94% RR)

CAS: 104934-50-1 | (C10H14S)n | NaN g/mol
製品番号(カタログ番号) H66726.ME
または、製品番号H66726-ME
価格(JPY)
-
数量:
500 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS104934-50-1
Molecular Formula(C10H14S)n
SMILESCCCCCCC1=C(-*)SC(-*)=C1
MDL NumberMFCD00217686
Appearance (Color)green to dark green to black
FormFlake or threadlike
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only