Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Fisher Chemical™

Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide na oh, soda lye, caustic soda, ascarite, white caustic, sodium hydrate, sodium hydroxide solution, soda, caustic, aetznatron, 6 ampules, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
数量:
1 Ampule
6 Ampules
製品番号(カタログ番号) J-7660C-05
または、製品番号J/7660C/05
価格(JPY)
-
お問い合わせください ›
数量:
1 Ampule
一括またはカスタム形式をリクエストする
仕様
沸点100°C
CAS1310-73-2
ChEBICHEBI:32145
Colorless
IUPAC名sodium;hydroxide
さらに表示
HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide na oh, soda lye, caustic soda, ascarite, white caustic, sodium hydrate, sodium hydroxide solution, soda, caustic, aetznatron, 6 ampules, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997