Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals

製品番号(カタログ番号)数量パッケージ
J17886.031 gPlastic bottle
J17886.065 gPlastic bottle
J17886.1425 gPlastic bottle
J17886.22100 gPlastic bottle
製品番号(カタログ番号) J17886.03
価格(JPY)
-
数量:
1 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS367-93-1
IUPAC Name(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
Molecular FormulaC9H18O5S
InChI KeyBPHPUYQFMNQIOC-NXRLNHOXSA-N
SMILESCC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
さらに表示
Optical Rotation-33° to -29° (c=1 in water)
Appearance (Color)White to pale cream
Assay from Suppliers CofA≥99% (HPLC)
Assay from Suppliers CofADioxane: ≤ 0.02% (GC)
Water Content (Karl Fischer Titration)≤ 2.0%
さらに表示
  • Induces the lac operon in E. coli by binding and inactivating the lac repressor.
  • Selection for lac Y mutants.
  • Selection for recombinant plasmids containing β-galactosidase fusion proteins.
  • Induces the cellular content of lactose permease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Induces lac operon in E. coli by binding and inactivating lac repressor
  • Selection for lac Y mutants
  • Selection for recombinant plasmids containing beta-galactosidase fusion proteins
  • Induces cellular content of lactose permease
RUO – Research Use Only