You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
Home
Shop All Products
Biochemical Reagents
Amino Acids
L-Leucine, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
Element Search
Structure Search
Thermo Scientific Chemicals
L-Leucine, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
Catalog number:
J18280.22
100 g, Plastic Bottle, Each
Certificates
Thermo Scientific Chemicals
L-Leucine, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
Catalog number:
J18280.22
100 g, Plastic Bottle, Each
Quantity
1 kg
100 g
5 kg
Packaging
Plastic Bottle
Certificates
Catalog number:
J18280.22
Price (USD)
Contact Us ›
Request bulk or custom quote
Save to list
Technical Support
Customer Service
Chemical Identifiers
CAS
61-90-5
IUPAC Name
(2S)-2-amino-4-methylpentanoic acid
Molecular Formula
C6H13NO2
InChI Key
ROHFNLRQFUQHCH-MLHKIVSYNA-N
SMILES
CC(C)C[C@H](N)C(O)=O
View More
Specifications
Specification Sheet
Assay (unspecified)
98.5-101.5% (dry basis)
Loss on Drying
≤ 0.2%
Thin Layer Chromatography
Related Compounds: Total Impurities: ≤ 2.0%
Residue on ignition
≤ 0.4%
Thin Layer Chromatography
Related Compounds: Individual Impurities: ≤ 0.5%
View More
Description
RUO – Research Use Only
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search