Thermo Scientific Chemicals

Thiabendazole, 98+%, Thermo Scientific Chemicals

Catalog number: J60009.09
10 g, Each
Thermo Scientific Chemicals

Thiabendazole, 98+%, Thermo Scientific Chemicals

Catalog number: J60009.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60009.09
also known as J60009-09
Price (USD)
Price: 31.80
Online price: 27.65
Your price:
Quantity
-

Chemical Identifiers

CAS
148-79-8
IUPAC Name
2-(1,3-thiazol-4-yl)-1H-1,3-benzodiazole
Molecular Formula
C10H7N3S
InChI Key
WJCNZQLZVWNLKY-UHFFFAOYSA-N
SMILES
N1C2=CC=CC=C2N=C1C1=CSC=N1
Heavy Metals
≤20 ppm
Form
Crystalline powder
Assay from Supplier's CofA
≥98.0%
Heavy Metals
≤20 ppm
Residue on ignition
≤0.5%

Description

Thiabendazole, a metal chelator and helminth-specific fumarate reductase inhibitor, is used primarily as an anthelmintic to control infections by helminth organisms. It is also used to control mold and fungal growth in plants. Also used to treat parasitic infections in humans. It is used clinically to treat threadworm, cutaneous larva migrans, visceral larva migrans, and trichinosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in methanol and dimethyl sulfoxide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text