Sodium bicarbonate, 1M buffer soln., pH 9.0
Sodium bicarbonate, 1M buffer soln., pH 9.0
Sodium bicarbonate, 1M buffer soln., pH 9.0
Thermo Scientific Chemicals

Sodium bicarbonate, 1M buffer soln., pH 9.0

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
数量:
250 mL
500 mL
製品番号(カタログ番号) J60066.AK
または、製品番号J60066-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
CAS144-55-8
ChEBICHEBI:32139
化学物質名または材質Sodium bicarbonate buffer soln.
IUPAC名sodium;hydrogen carbonate
InChIキーUIIMBOGNXHQVGW-UHFFFAOYSA-M
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only