HEPES, 1.0M buffer soln., pH 6.5, Sterile-Filtered
HEPES, 1.0M buffer soln., pH 6.5, Sterile-Filtered
HEPES, 1.0M buffer soln., pH 6.5, Sterile-Filtered
HEPES, 1.0M buffer soln., pH 6.5, Sterile-Filtered
Thermo Scientific Chemicals

HEPES, 1.0M buffer soln., pH 6.5, Sterile-Filtered

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
製品番号(カタログ番号)数量
J60111.AK
または、製品番号J60111-AK
250 mL
製品番号(カタログ番号) J60111.AK
または、製品番号J60111-AK
価格(JPY)
-
見積もりを依頼する
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
CAS7365-45-9
ChEBICHEBI:42334
化学物質名または材質HEPES buffer soln
Colorless
IUPAC名2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only