Citrate, 0.5M buffer soln., pH 5.0
Citrate, 0.5M buffer soln., pH 5.0
Citrate, 0.5M buffer soln., pH 5.0
Thermo Scientific Chemicals

Citrate, 0.5M buffer soln., pH 5.0

CAS: 6132-04-3294.10 g/mol
数量:
250 mL
500 mL
製品番号(カタログ番号) J60125.AK
または、製品番号J60125-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
CAS4-3-6132
Colorless
臭気Odorless
推奨保存方法Keep cold
濃度0.5M buffer soln.
さらに表示
Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
It is soluble in water (29.4 g/L) at 20°C. Insoluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. YP Lee.; T Takahashi. An improved colorimetric determination of amino acids with the use of ninhydrin. Analytical Biochemistry. 1972, 14,(1), 71-77.
  2. XZ Shu.; KJ Zhu.; W Song. Novel pH-sensitive citrate cross-linked chitosan film for drug controlled release. International Journal of Pharmaceutics. 2001, 212,(1), 19-28.