Thermo Scientific Chemicals

Pepstatin A, 98%

Catalog number: J60237.MB
25 mg, Protease Inhibitor, Each
Thermo Scientific Chemicals

Pepstatin A, 98%

Catalog number: J60237.MB
25 mg, Protease Inhibitor, Each
Quantity
Product Type
Catalog number: J60237.MB
also known as J60237-MB
Price (USD)
Price: 206.00
Online price: 175.65
Your price:
Quantity
-

Chemical Identifiers

CAS
26305-03-3
IUPAC Name
3-hydroxy-4-[(2S)-2-{3-hydroxy-6-methyl-4-[(2S)-3-methyl-2-[(2S)-3-methyl-2-(3-methylbutanamido)butanamido]butanamido]heptanamido}propanamido]-6-methylheptanoic acid
Molecular Formula
C34H63N5O9
InChI Key
FAXGPCHRFPCXOO-JHTFMCFMSA-N
SMILES
CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O
Appearance (Color)
White to cream
Form
Powder
Assay from Supplier's CofA
≥97.5% (HPLC)
Elemental Analysis
Carbon, C: 59.0-60.1%
Elemental Analysis
Hydrogen, H: 9.0-9.3%

Description

Pepstatin A, is used as an irreversible inhibitor of HIV-1, aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d. Pepstatin A also efficiently inhibits HIV-1 protease and Fcγ receptor induction by IFN-γ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text