Potassium phosphate, 0.5M buffer soln., pH 7.2
Potassium phosphate, 0.5M buffer soln., pH 7.2
Potassium phosphate, 0.5M buffer soln., pH 7.2
Thermo Scientific Chemicals

Potassium phosphate, 0.5M buffer soln., pH 7.2

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
数量:
250 mL
500 mL
製品番号(カタログ番号) J60326.AK
または、製品番号J60326-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
CAS7778-77-0
化学物質名または材質Potassium phosphate
IUPAC名potassium dihydrogen phosphate
InChIキーGNSKLFRGEWLPPA-UHFFFAOYSA-M
MDL番号MFCD00011401 MFCD00147253
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only