Trypsin, porcine pancreas
Trypsin, porcine pancreas
Trypsin, porcine pancreas
Thermo Scientific Chemicals

Trypsin, porcine pancreas

A pancreatic serine protease | CAS: 9002-07-7 | C35H47N7O10 | 23.8 kd g/mol
製品番号(カタログ番号) J60402.18
または、製品番号J60402-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9002-07-7
IUPAC Nameacetic acid; tert-butyl N-(1-{[1-({4-[(diaminomethylidene)amino]-1-[(5-methyl-2-oxo-2H-chromen-7-yl)carbamoyl]butyl}carbamoyl)-2-hydroxyethyl]carbamoyl}-2-phenylethyl)carbamate
Molecular FormulaC35H47N7O10
InChI KeyWGWZNYKOUXOZTC-UHFFFAOYNA-N
SMILESCC(O)=O.CC1=C2C=CC(=O)OC2=CC(NC(=O)C(CCCN=C(N)N)NC(=O)C(CO)NC(=O)C(CC2=CC=CC=C2)NC(=O)OC(C)(C)C)=C1
さらに表示
Assay (special)Trypsin activity: ≥ 250 USP units/mg
Appearance (Color)Light cream to cream
Assay (special)Chymotrypsin activity: ≤ 75 USP units/mg
Assay (special)E. coli, Salmonella, Pseudomonas aeruginosa: Absent in 25 gm
Assay (special)Endotoxin: 10 EU/mg
さらに表示
Trypsin, suitable for use in ELISA or for antigen applications in immunological protocols. Other applications include it is been used for the research purpose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reconstitute in aqueous buffer
RUO – Research Use Only
組成および保存条件
-30°C to -10°C