Sodium bicarbonate, 1M buffer soln., pH 8.5
Sodium bicarbonate, 1M buffer soln., pH 8.5
Sodium bicarbonate, 1M buffer soln., pH 8.5
Sodium bicarbonate, 1M buffer soln., pH 8.5
Thermo Scientific Chemicals

Sodium bicarbonate, 1M buffer soln., pH 8.5

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
数量:
250 mL
500 mL
製品番号(カタログ番号) J60408.AK
または、製品番号J60408-AK
価格(JPY)
-
見積もりを依頼する
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
CAS144-55-8
ChEBICHEBI:32139
化学物質名または材質Sodium bicarbonate buffer soln.
IUPAC名sodium;hydrogen carbonate
InChIキーUIIMBOGNXHQVGW-UHFFFAOYSA-M
さらに表示
Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong acids, strong oxidizing agents.
RUO – Research Use Only

General References:

  1. R Axéan.; S Ernback. Chemical fixation of enzymes to cyanogen halide activated polysaccharide carriers. European Journal of Biochemistry. 1971, 18 ,(3), 351-360.
  2. P Cuatrecasas.; I Parikh. Adsorbents for affinity chromatography. Use of N-hydroxysuccinimide esters of agarose. Biochemistry. 1972, 11 ,(12), 2291-2299.